Anti-FXR2 Mouse Monoclonal Antibody [clone: A42]

Supplier: GeneTex

GTX26085
89361-820EA 437.83 USD
89361-820
Anti-FXR2 Mouse Monoclonal Antibody [clone: A42]
Antibodies
Mouse Monoclonal antibody [A42] to FXR2

Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: FXR2
Clonality: Monoclonal
Clone: A42 
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR