Anti-FUZ Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX120464
89359-130EA 582.76 USD
89359-130
Anti-FUZ Rabbit Polyclonal Antibody
Antibodies
May be involved in cytoskeleton function and cell polarity in epithelial cells.

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: FUZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR