Anti-FTCD Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX109972
89356-128EA 557.66 USD
89356-128
Anti-FTCD Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to FTCD (formiminotransferase cyclodeaminase)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: FTCD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR