Anti-FSH Mouse Monoclonal Antibody [clone: INN-hFSH-6]

Supplier: GeneTex
GTX74590
89365-480EA 794.53 USD
89365-480
Anti-FSH Mouse Monoclonal Antibody [clone: INN-hFSH-6]
Antibodies
Mouse monoclonal antibody [INN-hFSH-6] to FSH beta

Type: Primary
Antigen: FSHB
Clonality: Monoclonal
Clone: INN-hFSH-6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR