Anti-FOXO3 Goat Polyclonal Antibody

Supplier: GeneTex
GTX17026
89360-262EA 577.91 USD
89360-262
Anti-FOXO3 Goat Polyclonal Antibody
Antibodies
FOXO3A belongs to the forkhead family of transcription factors which are characterized by a distinct forkhead domain. This gene likely functions as a trigger for apoptosis through expression of genes necessary for cell death. Translocation of this gene with the MLL gene is associated with secondary acute leukemia. Alternatively spliced transcript variants encoding the same protein have been observed.

Approx 90kDa band observed in human Brain and Heart lysates (calculated MW of 71.3kDa according to NP_001446 and NP_963853). Recommended concentration: 0.1-0.3µg/ml. Both variants represent identical protein (NP_001446.1and NP_963853.1).

Type: Primary
Antigen: FOXO3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR