Goat polyclonal antibody to FOXA1 / HNF3A
Approx 50kDa band observed in Human lysates of cell line HepG2 (calculated MW of 49.1kDa according to NP_004487.2). Recommended concentration: 0.2-0.6µg/ml. An additional band of unknown identity was also consistently observed at 35kDa. This band was successfully blocked by incubation with the immunising peptide. We would appreciate any feedback from people in the field - have any such results been reported with other antibodies/lysates? Have any further splice variants/modified forms been reported? Immunohistochemistry: In paraffin embedded Human Prostate shows nuclear staining of secretory cells. Recommended concentration, 5-10 µg/ml.
Type: Primary
Antigen: FOXA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse