Anti-NPFF Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX10352
89350-026EA 659.34 USD
89350-026
Anti-NPFF Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to FMRFamide

IHC-Fr: Use at a dilution of 1/1000 - 1/5000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: NPFF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR