Anti-FLUORESCEIN Goat Polyclonal Antibody

Supplier: GeneTex
GTX27253
89362-522EA 696.18 USD
89362-522
Anti-FLUORESCEIN Goat Polyclonal Antibody
Antibodies
Affinity purified polyclonal anti-fluorescein can be used to detect both fluoresceinated proteins and nucleic acids

Type: Primary
Antigen: FLUORESCEIN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR