Anti-FGFR4 Rabbit Polyclonal Antibody

GTX112934
89357-236EA 376.43 USD
89357-236
Anti-FGFR4 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to FGFR4 (fibroblast growth factor receptor 4)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: FGFR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR