Anti-FTL Mouse Monoclonal Antibody [clone: F31]

Supplier: GeneTex
GTX10061
89350-604EA 666.2 USD
89350-604
Anti-FTL Mouse Monoclonal Antibody [clone: F31]
Antibodies
Mouse monoclonal [F31] to Ferritin

ELISA: Use at an assay dependent dilution. Can be used with GTX10060 (capture). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: FTL
Clonality: Monoclonal
Clone: F31
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR