Anti-FTL Mouse Monoclonal Antibody [clone: 090-10175]

Supplier: GeneTex
GTX41016
89363-408EA 659.34 USD
89363-408
Anti-FTL Mouse Monoclonal Antibody [clone: 090-10175]
Antibodies
Mouse monoclonal [090-10175] to Ferritin

IA: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: FTL
Clonality: Monoclonal
Clone: 090-10175
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR