Anti-KDM2A Rabbit Polyclonal Antibody

GTX104869
89354-000EA 176.03 USD
89354-000
Anti-KDM2A Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to FBXL11 (F-box and leucine-rich repeat protein 11)

Antibody reactive against peptide, detects immunogen. Further validation in progress.

Type: Primary
Antigen: KDM2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR