Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FANCD2 Mouse Monoclonal Antibody [clone: FI-17]

Supplier: Genetex

GTX25360
89367-588EA 438.66 USD InStock
89367-588
Anti-FANCD2 Mouse Monoclonal Antibody [clone: FI-17]
Antibodies
Mouse monoclonal [FI - 17] to FANCD2

IP: Use at an assay dependent dilution. WB: Use at a dilution of 1/1000 - 1/2000. Detects a band of approximately 155 and 162 kDa (post-translationally modified form). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: FANCD2
Clonality: Monoclonal
Clone: FI-17
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty