Anti-FADD Rabbit Polyclonal Antibody

GTX73104
89353-124EA 376.85 USD
89353-124
Anti-FADD Rabbit Polyclonal Antibody
Rabbit Polyclonal to FADD ( FAS - Associated Death Domain - containing Protein )

For IHC-P: Use at a dilution of 1:2,000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: FADD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR