Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, Order Status will not be shown from 8:00am – 10:00am CEST Sunday.

Anti-ACTB Mouse Monoclonal Antibody [clone: NH3]

Supplier: Genetex

GTX76100
89365-988EA 212.63 USD InStock
89365-988
Anti-ACTB Mouse Monoclonal Antibody [clone: NH3]
Antibodies
Mouse monoclonal antibody [NH3] to F-Actin

Recommended Starting Dilutions: ELISA: Max Dilution 1 / 10 Flow Cytometry: Max Dilution 1 / 10 Western Blotting: 1/100 - 1/500 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: ACTB
Clonality: Monoclonal
Clone: NH3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse, Rat, Rabbit

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty