Anti-ETHE1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX115707
89358-390EA 690.85 USD
89358-390
Anti-ETHE1 Rabbit Polyclonal Antibody
Antibodies
This gene encodes a protein that is expressed in the thyroid. [provided by RefSeq]

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ETHE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR