Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ESRRA Rabbit Polyclonal Antibody

Supplier: Genetex
GTX12984
Estrogen-related receptor alpha (ERR alpha), a NR3 Steroid Receptor, was isolated based on sequence similarity in its DNA-binding domain to estrogen receptor alpha (ER alpha). ERR alpha has been shown to regulate the promoters of lactoferrin, medium-chain acyl CoA dehydrogenase, osteopontin, and thyroid receptor alpha, and it may affect cellular energy balance and bone formation. ERR alpha binds as a monodimer to the extended half-site TNAAGGTCA and as a homodimer to the estrogen response element (ERE) and is a constitutive activator of the estrogen response element and the palindromic thyroid hormone response element (TRE(pal)) but not of the glucocorticoid response element (GRE). ERR alpha 1 is the major isoform expressed in human breast cancer cell lines. Recent studies have shown that Phe-329 is responsible for the constitutive activity of ERR alpha. ERR alpha is a potential biomarker for unfavorable clinical outcome and, possibly, hormonal insensitivity in breast tumors. ERR alpha status may be predictive of sensitivity to hormonal blockade therapy, and ERR alpha status may also be predictive of ErbB2-based therapy such as Herceptin. Moreover, ERR alpha may be a candidate target for therapeutic development. ERRalpha null mice have altered regulation of genes involved in adipogenesis.In mouse, ERR alpha is expressed in many adult and embryonic tissues (particularly at the onset of ossification) as well as in several osteoblast cell lines. ERR alpha expression has been documented in mouse in brain, spinal cord, pituitary gland, heart, intestine, bone, brown adipose tissue, heart, uterus, cervix, nerve, skeletal muscle, and vagina. ESTs have been isolated from human tissue libraries, including cancerous blood, brain, breast, cervix, colon, duodenum, eye, head/neck, kidney, liver, lung, ovary, pancreas, skeletal muscle, skin, stomach, and uterus, and normal adrenal, blood, brain, colon, embryo, eye, head/neck, heart, kidney, prostate, skeletal muscle, testis, and uterus. The ligands for ERR alpha are PPARgamma coactivator 1 (PGC-1) beta and flavone and isoflavone phytoestrogens.

IHC-P: Use at a dilution of 1/100 - 1/200. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ESRRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
25μg GTX12984 89359-538 Each (25µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol ESRRA
Antigen Name ERR alpha
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Antigen Synonyms ERRa, ERRalpha, ERR1, NR3B1, ESRL1
Gene ID 2101
Concentration 1 mg/ml
Immunogen Synthetic peptide (unfortunately, the amino acid sequence is considered to be commercially sensitive)
Purification Immunogen affinity purified
Storage Buffer Preservative: 0.01% Sodium Azide; Constituents: PBS. pH 7.7
Storage Temperature Keep as concentrated solution. Store at 4ºC short term. For extended storage aliquot and store at -20ºC or below. Avoid freeze-thaw cycles.
Tested Applications IHC-P

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty