Anti-ERK5 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX25686
89361-708EA 611.89 USD
89361-708
Anti-ERK5 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to ERK5 (phospho Thr218/Tyr220)

IHC-Fr: Use at an assay dependent dilution. WB: Use at a dilution of 1/1000. Predicted molecular weight: 88 kDa. Due to the low abundance and low levels of activation of endogenous ERK5, over-expression or immunoprecipitation of ERK5 protein may be required to detect activation. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ERK5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR