Anti-ERBB3 Goat Polyclonal Antibody

Supplier: GeneTex
GTX89440
89366-988EA 607.67 USD
89366-988
Anti-ERBB3 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to ERBB3 / HER3

Approx 150kDa band observed in human lysates of T cell line JURKAT (calculated MW of 148kDa according to NP_001973.2). Recommended concentration: 0.1-0.3µg/ml. A minor band of unknown identity was also consistently observed at 48kDa. This band was successfully blocked by incubation with the immunising peptide. We would appreciate any feedback from people in the field - have any results been reported with other antibodies/lysates? Have any further splice variants/modified forms been reported? This antibody is expected to recognise isoform 1 (NP_001973.2) only.

Type: Primary
Antigen: ERBB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR