Anti-ERAB Mouse Monoclonal Antibody [clone: 10D12]

Supplier: GeneTex
GTX10259
89349-646EA 585.3 USD
89349-646
Anti-ERAB Mouse Monoclonal Antibody [clone: 10D12]
Antibodies
Mouse monoclonal [10D12] to ERAB - Mitochondrial Marker

Dot: Use at a dilution of 1/100 - 1/1000. IHC: Use at a dilution of 1/100 - 1/1000. WB: Use at a dilution of 1/100 - 1/1000. Predicted molecular weight: 27 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ERAB
Clonality: Monoclonal
Clone: 10D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR