Anti-EPHA10 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX85273
89366-816EA 628.47 USD
89366-816
Anti-EPHA10 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to EphA10

Type: Primary
Antigen: EPHA10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR