Anti-EPHB4 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX108595
89355-606EA 557.66 USD
89355-606
Anti-EPHB4 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to EphB4

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: EPHB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR