Mouse monoclonal antibody to p300 [RW128 ]
This antibody may be used in Western blot where a band is seen at ~300 kDa, representing the p300 protein. Past productions of this antibody have also been used for immunoprecipitation and immunofluorescence (formalin-fixed paraffin-embedded sections never tested) but this lot has not yet been tested in such applications. Suggested working dilutions:WB:1:250-1:500.ICC/IF:1:10-1:2000.IP:1:10-1:500.The investigator should determine the optimal working dilution for a specific application.
Type: Primary
Antigen: EP300
Clonality: Monoclonal
Clone: RW128
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human