Anti-Endothelium Mouse Monoclonal Antibody

Supplier: GeneTex
GTX76065
89365-978EA 678.11 USD
89365-978
Anti-Endothelium Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Endothelium
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR