Anti-ETRA Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX12978
89359-534EA 481.52 USD
89359-534
Anti-ETRA Rabbit Polyclonal Antibody
Antibodies
Receptor A for endothelin 1. Mediates its action by association with G proteins that activate a phosphatidylinositol calcium second messenger system.

Type: Primary
Antigen: ETRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR