Anti-COL18A1 Rabbit Polyclonal Antibody

GTX30875
89352-738EA 422.38 USD
89352-738
Anti-COL18A1 Rabbit Polyclonal Antibody
Rabbit polyclonal to Endostatin

For IHC-P, WB: Use at an assay dependent concentration. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: COL18A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR