Anti-Scnn1g Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX41972
89363-484EA 585.51 USD
89363-484
Anti-Scnn1g Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ENaC Gamma

We recommend a working dilution of 1:1000 (WB).

Type: Primary
Antigen: Scnn1g
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR