Anti-EIF2AK1 Rabbit Polyclonal Antibody

GTX111963
89356-938EA 376.76 USD
89356-938
Anti-EIF2AK1 Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to EIF2AK1 (eukaryotic translation initiation factor 2-alpha kinase 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: EIF2AK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR