Anti-EGFR Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX25636
89361-678EA 608.78 USD
89361-678
Anti-EGFR Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to EGFR (phospho Y845)

ICC: Use at an assay dependent dilution. WB: 1/1000. Detects a band of approximately 185 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: EGFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR