Anti-S1PR2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX87713
89366-944EA 590.12 USD
89366-944
Anti-S1PR2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to EDG5

We recommend the following starting dilutions: WB: 1:500~1:1000 ELISA: 1:40000Optimal dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: S1PR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR