Anti-ECT2 Goat Polyclonal Antibody

Supplier: GeneTex
GTX24863
89361-564EA 607.87 USD
89361-564
Anti-ECT2 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to ECT2

Approx 100kDa band reported in human U87-MG glioblastoma lysate at 2.5µg/ml. A 30kDa band of unknown identity was also observed.

Type: Primary
Antigen: ECT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR