Anti-EBV Mouse Monoclonal Antibody

Supplier: GeneTex
GTX73439
89353-176EA 609.32 USD
89353-176
Anti-EBV Mouse Monoclonal Antibody
Antibodies
This antibody is specific to 60 kD latent membrane protein (LMP-1) encoded by the BNLF1 gene of the EBV. Each clone reacts with different epitopes on the hydrophilic c-terminus of the cytoplasmic domain of LMP-1. This antibody stains strongly with EBV positive lymphoblastoid cell lines and EBV infected B cell immunoblasts in infectious mononucleosis.

Type: Primary
Antigen: EBV
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR