Anti-DYKDDDDK Chicken Polyclonal Antibody

Supplier: GeneTex
GTX85472
89353-940EA 605.27 USD
89353-940
Anti-DYKDDDDK Chicken Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DYKDDDDK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR