Anti-DUSP6A Chicken Polyclonal Antibody

Supplier: GeneTex
GTX19115
89367-500EA 663.11 USD
89367-500
Anti-DUSP6A Chicken Polyclonal Antibody
Antibodies
DUSP6a inactivates MAP kinases. It has a specificity for the ERK family, and is implicated in muscle and neuronal differentiation.

For WB: Use at an assay dependent dilution. Detects a band of approximately 60 kDa in an E-coli fusion protein sample (predicted molecular weight: 42 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: DUSP6A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR