Anti-DR5 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX24106
89367-576EA 616.35 USD
89367-576
Anti-DR5 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to DR5

For WB: Use at a dilution of 1:500 - 1:1,000. Not tested in other applications. Optimal concentrations/dilutions should be determined by the researcher.

Type: Primary
Antigen: DR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR