Anti-DP2 Mouse Monoclonal Antibody [clone: BC-2]

Supplier: GeneTex
GTX11835
89358-876EA 552.2 USD
89358-876
Anti-DP2 Mouse Monoclonal Antibody [clone: BC-2]
Antibodies
The metabolic function of the APC gene is not known. Defects in this gene cause familial adenomatous polyposis (FAP), an autosomal dominant pre-malignant disease that usually progresses to malignancy. Disease-associated mutations tend to be clustered in a small region designated the mutation cluster region (MCR) and result in a truncated protein product.

EMSA: Use at an assay dependent dilution. WB: 0.5-1?ug/mL using a whole extract of transfected 293T (human embryonal kidney) cells expressing DP2Not tested in other applications. IP: suitable using native and denatured DP2. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: DP2
Clonality: Monoclonal
Clone: BC-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR