Anti-IgA Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

GTX77076
89353-470EA 152.47 USD
89353-470
Anti-IgA Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
HRP-conjugated Goat anti-Dog IgA Polyclonal antibody

Recommended Starting Dilutions: WB: Use 1:2000-1:10,000. ELISA: Use 1:10,000 - 1:30,000. Optimal working dilutions should be determined for your particular assay conditions.

Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR