Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Polb Mouse Monoclonal Antibody [clone: 18S]

Supplier: Genetex

GTX23181
89351-914EA 358.58 USD InStock
89351-914
Anti-Polb Mouse Monoclonal Antibody [clone: 18S]
Antibodies
Mouse monoclonal [18S] to DNA Polymerase beta

Immunoprecipitation (denatured): use at a concentration of 2μg / mg. Western Blotting: use at a concentration of 1 - 2μg / ml for 2hrs at RT. Not tested in other applications. Optimal dilutions / concentrations should be determined by the end user. By Western blot, this antibody detects a band of 39 kDa, which corresponds to the predicted molecular weight of DNA Polymerase beta.

Type: Primary
Antigen: Polb
Clonality: Monoclonal
Clone: 18S
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Hamster, Bovine

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty