Anti-DLL4 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX109649
89355-972EA 427.1 USD
89355-972
Anti-DLL4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Delta-4 (delta-like 4 (Drosophila))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: DLL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR