Anti-Digoxin Mouse Monoclonal Antibody

Supplier: GeneTex
GTX40814
89363-322EA 659.34 USD
89363-322
Anti-Digoxin Mouse Monoclonal Antibody
Antibodies
Digoxin is a beta blocker drug used to treat heart problems, such as heart failure, high blood pressure.

Type: Primary
Antigen: Digoxin
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR