Anti-DGAT1 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX48577
89363-666EA 304.56 USD
89363-666
Anti-DGAT1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to DGAT1

Western Blot 2.0 ug/ ml

Type: Primary
Antigen: DGAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR