Anti-DDDDK TAG Goat Polyclonal Antibody

Supplier: GeneTex
GTX21259
89351-620EA 554.79 USD
89351-620
Anti-DDDDK TAG Goat Polyclonal Antibody
Antibodies
This is a useful tool for the localization and characterization of DDDDK-tagged proteins.

Type: Primary
Antigen: DDDDK TAG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR