Anti-DDAH2 Goat Polyclonal Antibody

Supplier: GeneTex
GTX21383
89361-016EA 617.52 USD
89361-016
Anti-DDAH2 Goat Polyclonal Antibody
Antibodies
This gene belongs to the dimethylarginine dimethylaminohydrolase (DDAH) gene family. The encoded enzyme plays a role in nitric oxide generation by regulating cellular concentrations of methylarginines, which in turn inhibit nitric oxide synthase activity. [provided by RefSeq]

Western Blot - Approx 40-45kDa band seen in Mouse Heart extracts. Recommended for use at 1-3?g/ml.Please note that DDAH2, according to NP_039268, has a consensus glycosylation site. Glycosylation can affect migration of proteins on SDS-PAGE [Hames, B.D and Rickwood, D., eds. Gel electrophoresis of proteins: a practical approach. Oxford University Press]. This may be the reason why we see an approx 40kDa band rather than the predicted 33 kDa band.

Type: Primary
Antigen: DDAH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR