Anti-CK5 Mouse Monoclonal Antibody [clone: LH8]

Supplier: GeneTex
GTX40203
89363-130EA 602.81 USD
89363-130
Anti-CK5 Mouse Monoclonal Antibody [clone: LH8]
Antibodies
Mouse monoclonal [LH8] to Cytokeratin 5+14

IHC: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CK5
Clonality: Monoclonal
Clone: LH8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR