Anti-MAPT Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX24841
89361-542EA 608.78 USD
89361-542
Anti-MAPT Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Tau ( phospho T205 )

For WB: Use at a dilution of 1:1000. Predicted molecular weight: 62 kDa. Can be blocked with GTX25239 Tau peptide - phospho T205 (phospho and non-phospho pair). Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MAPT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR