Anti-MAPT Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX24861
89361-560EA 611.82 USD
89361-560
Anti-MAPT Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Tau ( phospho S409 )

WB: Use at a dilution of 1/1000. Predicted molecular weight: 62 kDa. Can be blocked with GTX25228 Tau peptide - phospho S409 (phospho and non-phospho pair) . Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MAPT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR