Anti-MAPT Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX24856
89361-552EA 589.82 USD
89361-552
Anti-MAPT Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Tau ( phospho S262 )

For WB: Use at a dilution of 1:1000. Predicted molecular weight: 62 kDa. Can be blocked with GTX25234 Tau peptide - phospho S262 (phospho and non-phospho pair) . Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MAPT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR