Anti-RPL7 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX87592
89366-930EA 616.68 USD
89366-930
Anti-RPL7 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to RPL7

We recommend the following starting dilutions: WB: 1:500~1:1000 ELISA: 1:10000Optimal dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: RPL7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR