Anti-ROBO1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX36928
89363-036EA 750.48 USD
89363-036
Anti-ROBO1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Robo1

For ELISA: Use at a dilution of 1:30,000-1:160,000. For IF: Use at an assay dependent dilution. For IHC-P: Use at a concentration of 2-10?g/ml. For IP: Use at an assay dependent dilution. For WB: Use at a dilution of 1:500-1:3,000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ROBO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR