Anti-ROBO1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX89308
89366-982EA 607.87 USD
89366-982
Anti-ROBO1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to ROBO1 / DUTT1 (Internal)

Approx 250kDa band observed in Human Endothelial Cell line HUVEC lysates (calculated MW of 181kDa according to NP_002932.1). Some of the observed proteolytic breakdown products are also found in Human, Mouse and Rat Brain lysates. Recommended concentration: 1-3µg/ml. This antibody is expected to recognise all reported isoforms (NP_002932.1; NP_598334.1; NP_001139316.1; NP_001139317.1).

Type: Primary
Antigen: ROBO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR